Learn More
Biomatik Corporation Human Apelin 12 (AP12) CLIA Kit
Supplier:  Biomatik Corporation EKU08645
Encompass_Preferred
Human Apelin 12 (AP12) CLIA Kit for detecting AP12 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 62.5-16,000pg/ml. Sensitivity: 22.8pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Chemiluminescence. Assay Time: 2h. Accession Number: Q9ULZ1. Gene ID: 8862. Target Synonyms: APLN12. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6509