Learn More
Biomatik Corporation Human Apelin 12 (AP12) ELISA Kit
Supplier:  Biomatik Corporation EKU02472
Encompass_Preferred
Human Apelin 12 (AP12) ELISA Kit for detecting AP12 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 74.07-6,000pg/ml. Sensitivity: 25.64pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: Q9ULZ1. Gene ID: 8862. Target Synonyms: APLN12. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5911