Learn More
Raybiotech Inc Human/Mouse/Rat Apelin EIA Kit.
Supplier:  Raybiotech Inc EIAAPC1
Encompass_Preferred
Optimized for serum and cell culture medium. Competition-based ELISA on a 96-well strip plate.
Catalog No. 50-194-8799