Learn More
Abcam Human APLP1 ELISA Kit - Extracellular, 1 x 96 Tests
Catalog No. 502669937
Encompass_Preferred
Human APLP1 ELISA Kit - Extracellular is a single-wash 90-min Simplestep used to quantify Human APLP1 - Extracellular with a sensitivity of 7.5 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Validated on a number of sample types including cerebrospinal fluid (CSF) - Design your own immunoassay: we also offer the conjugation-ready antibody pair
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-266-9937 Supplier Abcam Supplier No. AB3091121X96TESTS
Human APLP1 ELISA Kit - Extracellular is a single-wash 90-min Simplestep used to quantify Human APLP1 - Extracellular with a sensitivity of 7.5 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Validated on a number of sample types including cerebrospinal fluid (CSF) - Design your own immunoassay: we also offer the conjugation-ready antibody pair
The product is subject to the following: Abcam Restricted Use Statement