Learn More
ILEX LIFE SCIENCES LLC Apo Transferrin Human, Native Protein (Serum), 500 mg
SDP
Catalog No. 502339522
Encompass_Preferred
Apo Transferrin Human, Native Protein (Serum) is a glycoprotein purified from human serum that has a molecular mass of approximately 77 kDa. It has a low iron content of ~9 ppm. FORMULATION: Sterile filtered white lyophilized (freeze-dried) powder. The protein (10 mg/ml) was lyophilized from 20mM NH4HC03 solution. May contain traces of buffer salts. PURITY: Greater than 98.0%. IRON CONTENT: The iron content was estimated by ICP and was found to be 9 ppm. SAFETY: Plasma from each donor has been tested and found negative for antibody to HIV-1, HIV-2, HCV, HBSAG and Syphilis. OTHER NAMES: Human ApoTransferrin, iron-free transferrin, Serotransferrin, Transferrin, Siderophilin, Beta-1-metal-binding globulin, TF, PRO1557, PRO2086, DKFZp781D0156, ATF.
Catalog No. 50-233-9522 Supplier ILEX LIFE SCIENCES LLC Supplier No. A13250500
Apo Transferrin Human, Native Protein (Serum) is a glycoprotein purified from human serum that has a molecular mass of approximately 77 kDa. It has a low iron content of ~9 ppm. FORMULATION: Sterile filtered white lyophilized (freeze-dried) powder. The protein (10 mg/ml) was lyophilized from 20mM NH4HC03 solution. May contain traces of buffer salts. PURITY: Greater than 98.0%. IRON CONTENT: The iron content was estimated by ICP and was found to be 9 ppm. SAFETY: Plasma from each donor has been tested and found negative for antibody to HIV-1, HIV-2, HCV, HBSAG and Syphilis. OTHER NAMES: Human ApoTransferrin, iron-free transferrin, Serotransferrin, Transferrin, Siderophilin, Beta-1-metal-binding globulin, TF, PRO1557, PRO2086, DKFZp781D0156, ATF.