Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human ApoA-I Recombinant Protein, PeproTech®

Catalog No. 35011100UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
500 μg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
35011100UG 100 μg
350-11-1MG 1 mg
350-11-20UG 20 μg
35011250UG 250 μg
35011500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 35011100UG Supplier Gibco™ Supplier No. 35011100UG
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human ApoA-I is a 28.2 kDa protein of 244 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Apolipoprotein A-1 (apo A-1) in human plasma is a major protein component of high density lipoprotein (HDL). The molecular mass is 24 kDa. Apo A-1 is a cofactor for enzymes involved in the normal turnover of cholesterol and is produced in the liver. Plasma concentration is 270-380 mg/100 mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P02647
For Use With (Application) Control, Western Blot Control, Western Blot
Formulation Protein with no preservative
Gene ID (Entrez) 335
Molecular Weight (g/mol) 28.2 kDa
Name Human ApoA-I
Purification Method Purified
Quantity 100 μg
Source E. coli
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias 1-Sep; 2-Sep; Alp-1; Apo a; APO A1; apo A-1; APO I; apo(a); APOA1; Apoa-1; ApoAI; Apo-AI; apoA-I; Apolipoprotein; Apolipoprotein A1; apolipoprotein A-1; Apolipoprotein AI; apolipoprotein A-I; apolipoprotein A-I precursor; Apolipoprotein A-I(1-242); Brp-14; Ltw-1; Lvtw-1; MGC102525; MGC117399; MGC124929; preproapolipoprotein A-I; prepro-apolipoprotein-AI; ProapoA-I; Proapolipoprotein A-I; Sep-1; Sep2; Sep-2; Truncated apolipoprotein A-I; Udpgt; Ugt1; Ugt1a7
Common Name ApoA1
Gene Symbol APOA1
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence MDEPPQSPWD RVKDLATVYV DVLKDSGRDY VSQFEGSALG KQLNLKLLDN WDSVTSTFSK LREQLGPVTQ EFWDNLEKET EGLRQEMSKD LEEVKAKVQP YLDDFQKKWQ EEMELYRQKV EPLRAELQEG ARQKLHELQE KLSPLGEEMR DRARAHVDAL RTHLAPYSDE LRQRLAARLE ALKENGGARL AEYHAKATEH LSTLSEKAKP ALEDLRQGLL PVLESFKVSF LSALEEYTKK LNTQ
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 97% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.