Learn More
Sino Biological Recombinant Human Apolipoprotein A-I/APOA1 Protein (His Tag) 1 mg
Supplier:  Sino Biological 10686H07E1
Encompass_Preferred
A DNA sequence encoding the mature form of human APOA1 (P02647) (Asp25-Gln267) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-224-7184