Learn More
Proteintech Group Inc Human APOA1 ELISA Kit 96well
SDP
Supplier:  Proteintech Group Inc KE0015796T
Encompass_Preferred
APOA1 (apolipoprotein A1) is a plasma protein and a major protein component of high density lipoproteins (HDL) which is associated with reversed cholesterol transport, lipid/cholesterol binding, lecithin/cholesterol acyltransferase (LCAT) activation and specific receptors binding. It is synthesized in the liver and small intestine. Defects of APOA1 cause low HDL level and systemic non-neuropathic amyloidosis. Serum concentration of APOA1 is inversely related to the risk of developing atherosclerosis. Alterations of APOA1 level also correlate to cancer and Alzheimer's Disease, thus measurement of APOA1 can be used in diagnosis or prognosis of related diseases.
Catalog No. 50-197-4112