Learn More
Sino Biological Human APOA1 ELISA Pair Set 5PP (Sino Biological)
Catalog No. 502372483
Encompass_Preferred
Each vial contains 278 ng of recombinant Human Apolipoprotein A-I / ApoA1. Reconstitute with 1 mL dilution buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A six-point standard curve using 2-fold serial dilutions in dilution buffer, and a high standard of 12000 pg/mL is recommended.
Catalog No. 50-237-2483 Supplier Sino Biological Supplier No. SEK106865
Each vial contains 278 ng of recombinant Human Apolipoprotein A-I / ApoA1. Reconstitute with 1 mL dilution buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A six-point standard curve using 2-fold serial dilutions in dilution buffer, and a high standard of 12000 pg/mL is recommended.