Learn More
Sino Biological Human APOA1 MAb 100ul (Sino Biological)
Supplier:  Sino Biological 10686MM09100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Apolipoprotein A-I/APOA1 (rh Apolipoprotein A-I/APOA1; Catalog#10686-H07E; P02647; Asp25-Gln267). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-3494