Learn More
Sino Biological Recombinant Anti-Apolipoprotein A-I/APOA1 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10686R03120
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human Apolipoprotein A-I / ApoA1 (rh Apolipoprotein A-I / ApoA1; Catalog#10686-H07E; P02647; Asp25-Gln267).
Catalog No. 50-240-8082