Learn More
Biomatik Corporation Human APOA1 (Apolipoprotein A-I) ELISA Kit
Supplier:  Biomatik Corporation EKF58494
Encompass_Preferred
Human APOA1 (Apolipoprotein A-I) ELISA Kit for detecting APOA1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P02647. Target Synonyms: APOA1; Apo-A1; Alp-1; Brp-14; Ltw-1; Lvtw-1; Sep-1; Sep-2; apo-AI; Apo-AI; ApoA-I; apoA-I; Apolipoprotein A1; apolipoprotein A-I; MGC117399. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9175