Learn More
BIOHIPPO Human APOA1BP Microsample
SDP
Catalog No. 5000315414
Encompass
Human APOA1BP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of APOA1BP in serum plasma urine tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 06 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human APOA1BP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human APOA1BP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human APOA1BP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15414 Supplier BIOHIPPO Supplier No. BHE15211409
Human APOA1BP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of APOA1BP in serum plasma urine tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 06 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human APOA1BP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human APOA1BP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human APOA1BP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color