Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human ApoA2 ELISA Kit

Catalog No. EH33RBX10
Encompass_Preferred
Change view
Click to view available options
10 x 96 Tests; HRP
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
EH33RBX10 10 x 96 Tests; HRP
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EH33RBX10 Supplier Invitrogen™ Supplier No. EH33RBX10
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

ELISA

Human ApoA2 quantitates human ApoA2 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human ApoA2.

At least 9 distinct polymorphic forms of apolipoproteins are known. The apolipoproteins act as stabilizers of the intact lipoprotein particles. Quantitative measurements of HDL, LDL, and VLDL particles of human serum are often used to estimate an individual's relative risk of coronary heart disease. In addition, quantitative immunological measurements of certain apolipoproteins ( especially A-1 and B) have been suggested to be more accurate estimators of coronary heart disease that measurements of lipoprotein particles (especially HDL and LDL). The apolipoproteins are derived from human plasma by density gradient ultracentrifugation and HPLC.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P02652
Assay Range 0.025-20 ng/mL
Assay Sensitivity 0.025 ng/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias Alp-2; APO A2; APOA2; Apoa-2; ApoAII; Apo-AII; ApoA-II; Apolipoprotein; Apolipoprotein A2; Apolipoprotein AII; apolipoprotein A-II; Apolipoprotein A-II(1-76); Hdl-1; ProapoA-II; Proapolipoprotein A-II; Truncated apolipoprotein A-IIAlp-2; APO A2; APOA2; Apoa-2; ApoAII; Apo-AII; ApoA-II; Apolipoprotein; Apolipoprotein A2; Apolipoprotein AII; apolipoprotein A-II; Apolipoprotein A-II(1-76); Hdl-1; ProapoA-II; Proapolipoprotein A-II; Truncated apolipoprotein A-II;
Gene ID (Entrez) 336
Gene Symbol APOA2
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 10 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target ApoA2
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.