Learn More
IBL America Apolipoprotein B-100 (ApoB-100)(Human) ELISA
Supplier:  IBL America 27181
Encompass_Preferred
Solid phase sandwich ELISA for the determination of Human ApoB-100 in serum, EDTA-plasma and cell culture media. For research use only, not for use in diagnostic procedures.
Catalog No. 50-212-0310