Learn More
Proteintech Group Inc Human APOB ELISA Kit 96well
SDP
Supplier:  Proteintech Group Inc KE0015896T
Encompass_Preferred
The apolipoprotein B (APOB) is a plasma protein synthesized primarily in the liver and intestine and play an important role in lipid and cholesterol metabolism. The APOB gene encodes two different isoproteins through mRNA editing, APOB-48 and APOB-100. APOB-100 is synthesized exclusively by the liver and is essential for the assembly of VLDL in the liver. APOB-48 is synthesized exclusively by the small intestine and is essential for the assembly of chylomicrons in the intestine. APOB-48 and APOB-100 share a common N-terminal sequence, but APOB-48 lacks APOB-100's C-terminal LDL receptor binding region. Plasma APOB levels are associated with coronary heart disease.
Catalog No. 50-197-4113