Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human APOB native Protein from human plasma

Catalog No. 89935306 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-935-306 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-935-306 Supplier Abnova™ Supplier No. P5328
Add to Cart
Add to Cart

Used for SDS-PAGE

This gene product is the main apolipoprotein of chylomicrons and low density lipoproteins. It occurs in plasma as two main isoforms, apoB-48 and apoB-100: the former is synthesized exclusively in the gut and the latter in the liver. The intestinal and the hepatic forms of apoB are encoded by a single gene from a single, very long mRNA. The two isoforms share a common N-terminal sequence. The shorter apoB-48 protein is produced after RNA editing of the apoB-100 transcript at residue 2180 (CAA->UAA), resulting in the creation of a stop codon, and early translation termination. Mutations in this gene or its regulatory region cause hypobetalipoproteinemia, normotriglyceridemic hypobetalipoproteinemia, and hypercholesterolemia due to ligand-defective apoB, diseases affecting plasma cholesterol and apoB levels. [provided by RefSeq]

Specifications

For Use With (Application) SDS-PAGE
Formulation Lyophilized from 1 ml of 10mM sodium deoxycholate, 50mM NaCl, 5 0mM Na
Gene ID (Entrez) 338
Name APOB (Human) Native Protein
Preparation Method Purified from low density lipoproteins (LDL) isolated by density gradient ultracentrifugation (1.03 - 1.05 g/mL density range) and by gel filtration chromatography in 10 mM sodium deoxycholate, 50 mM NaCl, 50 mM Na2CO3, pH 10.1.
Purification Method Density gradient ultracentrifugation and gel filtration chromatography
Quantity 500 μg
Storage Requirements Store at -20°C or lower on dry atmosphere. After reconstitution, store at 2-8°C no longer than 4 weeks. Reconstitution: Dissolve lyophilized Apo B in 100 mL to 1 mL of distilled or deionized water. If dissolving in solvent we recommend using 10 mM sodium deoxycholate, 50 mM NaCl, 50 mM Na2CO3, pH 10 to keep the sodium deoxycholate at an acceptable level instead of water.
Regulatory Status RUO
Gene Alias FLDB
Common Name APOB
Gene Symbol APOB
Species Human
Protein Tag None
Expression System Purified from low density lipoproteins (LDL) isolated by density gradient ultracentrifugation (1.03 - 1.05 g/mL density range) and by gel filtration chromatography in 10 mM sodium deoxycholate, 50 mM NaCl, 50 mM Na2CO3, pH 10.1.
Form Lyophilized
Purity or Quality Grade ≥95% by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.