Learn More
Raybiotech Inc Human ApoC1 ELISA Kit
Supplier:  Raybiotech Inc ELHAPOC11
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 3 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects human Apo C1. Other species not determined.
Catalog No. 50-194-8992