Learn More
Sigma Aldrich Fine Chemicals Biosciences Human APOC3 Apolipoprote
Catalog No. 502906516
Encompass
The antibody pair provided in this kit recognizes Human Apolipoprotein C3.ApoC-III is a glycoprotein that has 79 amino acids. It is coded by APOC3 gene. APOC3 is expressed in the liver and intestine. This gene is located on human chromosome 11.
Catalog No. 50-290-6516 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. RAB06181KT
The antibody pair provided in this kit recognizes Human Apolipoprotein C3.ApoC-III is a glycoprotein that has 79 amino acids. It is coded by APOC3 gene. APOC3 is expressed in the liver and intestine. This gene is located on human chromosome 11.