Learn More
Sigma Aldrich Fine Chemicals Biosciences Human APOE Apolipoprotei
Catalog No. 502906507
Encompass
Human APOE / Apolipoprotein E ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of apolipoprotein E in biological samples such as a serum plasma cell culture supernatants urine and/or cell and tissue lysates. APO-E plays an important role in the synthesis of chylomicrons and very-low-density lipoprotein (VLDL).
Catalog No. 50-290-6507 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. RAB06131KT
Human APOE / Apolipoprotein E ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of apolipoprotein E in biological samples such as a serum plasma cell culture supernatants urine and/or cell and tissue lysates. APO-E plays an important role in the synthesis of chylomicrons and very-low-density lipoprotein (VLDL).