Learn More
Raybiotech Inc Human ApoE ELISA Kit
Supplier:  Raybiotech Inc ELHAPOE1
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 1.5 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects human ApoE (isoforms ApoE2, ApoE3 and ApoE4).
Catalog No. 50-194-8995