Learn More
Bon Opus Biosciences Recombinant Human Apolipoprotein E/ApoE (C-6His) 1MG
SDP
Supplier:  Bon Opus Biosciences CI021MG
Encompass_Preferred
Recombinant Human Apolipoprotein E is produced by our Mammalian expression system and the target gene encoding Lys19-His317 is expressed with a 6His tag at the C-terminus. Accession# P02649. Formulation: Lyophilized from a 0.2 μm filtered solution of 20mM PB,150mM NaCl,pH7.4. Bon Opus Cat# CI02
Catalog No. 50-217-2797