Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human ApoH ELISA Kit

Catalog No. EH36RBX5
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH36RBX5 5 x 96 Tests
EH36RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EH36RBX5 Supplier Invitrogen™ Supplier No. EH36RBX5
Get Up to $3,000 in Extra Life Sciences Consumables with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ELISA

Human ApoH quantitates human ApoH in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human ApoH.

Apolipoprotein H has been implicated in a variety of physiologic pathways including lipoprotein metabolism, coagulation, and the production of antiphospholipid autoantibodies. APOH may be a required cofactor for anionic phospholipid binding by the antiphospholipid autoantibodies found in sera of many patients with lupus and primary antiphospholipid syndrome, but it does not seem to be required for the reactivity of antiphospholipid autoantibodies associated with infections.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P02749
Assay Range 30-6000 pg/mL
Assay Sensitivity 30 pg/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias 2GPI; activated protein C-binding protein; Anticardiolipin cofactor; APC inhibitor; APOH; Apo-H; Apolipoprotein H; apolipoprotein H (beta-2-glycoprotein I); B2G1; B2GP1; B2GPI; beta(2)GPI; beta-2-glycoprotein 1; Beta-2-glycoprotein I; beta-2-GPI; beta2-GPI; BG2GPI; activated protein C-binding protein; Anticardiolipin cofactor; APC inhibitor; APOH; Apo-H; Apolipoprotein H; apolipoprotein H (beta-2-glycoprotein I); B2G1; B2GP1; B2GPI; beta(2)GPI; beta-2-glycoprotein 1; Beta-2-glycoprotein I; beta-2-GPI; beta2-GPI; BG;
Gene ID (Entrez) 350
Gene Symbol APOH
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target Apolipoprotein H
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.