Learn More
Sino Biological Human APOH ELISA Pair Set 15PP (Sino Biological)
Catalog No. 502371756
Encompass_Preferred
Each vial contains 45 ng of recombinant Human Apolipoprotein H / APOH. Reconstitute with 1 mL dilution buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in dilution buffer, and a high standard of 4000 pg/mL is recommended.
Catalog No. 50-237-1756 Supplier Sino Biological Supplier No. SEK1122115
Each vial contains 45 ng of recombinant Human Apolipoprotein H / APOH. Reconstitute with 1 mL dilution buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in dilution buffer, and a high standard of 4000 pg/mL is recommended.