Learn More
Sino Biological Anti-Apolipoprotein H Antibody, Mouse Monoclonal
Supplier:  Sino Biological 11221MM0820
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Apolipoprotein H (rh Apolipoprotein H; Catalog#11221-H08H; NP_000033.2; Met1-Cys345). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-240-8570