Learn More
Sino Biological Human APOH MAb 100ul (Sino Biological)
Supplier:  Sino Biological 11221MM06100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human APOH (rh APOH; Catalog#11221-H08H; NP_000033.2; Met 1-Cys 345). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-3826