Learn More
Sino Biological Anti-Apolipoprotein H Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 201112T08100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human Apolipoprotein H / APOH fragment, and purified by antigen affinity chromatography.
Catalog No. 50-241-1741