Learn More
Sino Biological Human APOH PAb 100ul (Sino Biological)
Supplier:  Sino Biological 11221RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human APOH (rh APOH; Catalog#11221-H08H; NP_000033.2; Met 1-Cys 345). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-237-2974