Learn More
Biomatik Corporation Human Apolipoprotein A4 (APOA4) CLIA Kit
Supplier:  Biomatik Corporation EKU09778
Encompass_Preferred
Human Apolipoprotein A4 (APOA4) CLIA Kit for detecting APOA4 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.21-150ng/ml. Sensitivity: 0.08ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P06727. Gene ID: 337. Target Synonyms: ApoA-IV; ApoAIV; Apo-A4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8437