Learn More
Biomatik Corporation Human Apolipoprotein B100 (APOB100) ELISA Kit
Supplier:  Biomatik Corporation EKU02503
Encompass_Preferred
Human Apolipoprotein B100 (APOB100) ELISA Kit for detecting APOB100 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 93.75-6,000ng/ml. Sensitivity: 36.72ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P04114. Gene ID: 338. Target Synonyms: Apo-B100. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5960