Learn More
Biomatik Corporation Human Apolipoprotein B48 (APOB48) CLIA Kit
Supplier:  Biomatik Corporation EKU09792
Encompass_Preferred
Human Apolipoprotein B48 (APOB48) CLIA Kit for detecting APOB48 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.69-500pg/ml. Sensitivity: 0.28pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P04114. Gene ID: 338. Target Synonyms: Apo-B48. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8432