Learn More
Biomatik Corporation Human Apolipoprotein C4 (APOC4) ELISA Kit
Supplier:  Biomatik Corporation EKU02514
Encompass_Preferred
Human Apolipoprotein C4 (APOC4) ELISA Kit for detecting APOC4 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.64ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P55056. Gene ID: 346. Target Synonyms: Apo-C4; Apolipoprotein C-IV. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6918