Learn More
ACROBiosystems ACROBiosystems Human Apolipoprotein E / APOE Protein (R154S,R176C), His Tag
Catalog No. 502106481
Encompass_Preferred
Recombinant Protein;100UG;Human Apolipoprotein E (R154S,R176C), His Tag (APE-H5256) is expressed from human 293 cells (HEK293). It contains AA Lys 19 - His 317 (R154S,R176C) (Accession # P02649-1 (R154S,R176C)).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 36.1 kDa. The protein migrates as 35-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-6481 Supplier ACROBiosystems Supplier No. APEH5256100UG
Recombinant Protein;100UG;Human Apolipoprotein E (R154S,R176C), His Tag (APE-H5256) is expressed from human 293 cells (HEK293). It contains AA Lys 19 - His 317 (R154S,R176C) (Accession # P02649-1 (R154S,R176C)).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 36.1 kDa. The protein migrates as 35-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.