Learn More
Biomatik Corporation Human Apolipoprotein E (APOE) CLIA Kit
Supplier:  Biomatik Corporation EKU08911
Encompass_Preferred
Human Apolipoprotein E (APOE) CLIA Kit for detecting APOE in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.82-600ng/ml. Sensitivity: 0.37ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P02649. Gene ID: 348. Target Synonyms: Apo-E; AD2; Apoprotein; Alzheimer Disease 2 (E4-Associated,Late Onset. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7177