Learn More
Biomatik Corporation Human Apolipoprotein H (APOH) ELISA Kit
Supplier:  Biomatik Corporation EKU02524
Encompass_Preferred
Human Apolipoprotein H (APOH) ELISA Kit for detecting APOH in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.143ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P02749. Gene ID: 350. Target Synonyms: Apo-H; B2G1; BG; B2-GP1; B2GP1; Autoantigen β2 GP I; Previously; Beta 2 Glycoprotein 1; APC inhibitor; Activated protein C-binding protein; Anticardiolipin cofactor; Beta-2-GPI. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9123