Learn More
Description
RP-43146 contains native human b2-Glycoprotein I protein. RP-43146 may be used as a positive control in many immunological assays. The product is of human blood/plasma origin. Although the starting material was tested prior to initiation of the manufacturing process, and was found negative or nonreactive for anti-HIV-1/2, HIV-1 antigen(s), HBsAg, STS, anti-HCV, anti-HBcore and anti-HTLV I & II, extreme caution should be used when handling this material as there is a margin of error in all tests. This product is to be used for IN VITRO research purposes only, and is not intended for clinical or diagnostic use! Reconstitute with 1.02 mL of distilled water to achieve 0.1 mg/mL.
Specifications
Specifications
| Accession Number | P02749 |
| Concentration | 0.1 mg/mL |
| For Use With (Application) | Neutralization |
| Formulation | Protein with 0.2M glycine, 0.15M NaCl and no preservative; pH 7.4 |
| Gene ID (Entrez) | 350 |
| Molecular Weight (g/mol) | 54.2 kDa |
| Name | Human Apolipoprotein H |
| pH Range | 7.4 |
| Purification Method | SDS-PAGE |
| Quantity | 100 μg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.