Learn More
Biomatik Corporation Human Apolipoprotein L Domain Containing Protein 1 (APOLD1) ELISA Kit
Supplier:  Biomatik Corporation EKU12093
Encompass_Preferred
Human Apolipoprotein L Domain Containing Protein 1 (APOLD1) ELISA Kit for detecting APOLD1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.112ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q96LR9. Gene ID: 81575. Target Synonyms: VERGE; Vascular early response gene protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6279