Learn More
Biomatik Corporation Human Apolipoprotein L2 (APOL2) ELISA Kit
Supplier:  Biomatik Corporation EKU02529
Encompass_Preferred
Human Apolipoprotein L2 (APOL2) ELISA Kit for detecting APOL2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.062ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9BQE5. Gene ID: 23780. Target Synonyms: Apo-L2; APOL-II. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9121