Learn More
BIOHIPPO Human APOO Microsample
SDP
Catalog No. 5000315045
Encompass
Human APOO Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of APOO in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 051 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human APOO Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human APOO Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human APOO biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15045 Supplier BIOHIPPO Supplier No. BHE15211587
Human APOO Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of APOO in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 051 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human APOO Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human APOO Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human APOO biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color