Learn More
Raybiotech Inc C-Series Human Apoptosis Antibody Array 1 Kit.
Catalog No. 501948477
Encompass_Preferred
Detects 43 Human Apoptotic Factors. Suitable for all liquid sample types but intended for use with cell and tissue lysates. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: bad, bax, bcl-2, bcl-w, BID, BIM, Caspase-3, Caspase-8, CD40 (TNFRSF5), CD40 Ligand (TNFSF5), cIAP-2, Cytochrome C, DR6 (TNFRSF21), Fas (TNFRSF6/Apo-1), Fas Ligand (TNFSF6), HSP27, HSP60, HSP70, HTRA2, IGF-1, IGF-2, IGFBP-1, IGFBP-2, IGFBP-3, IGFBP-4, IGFBP-5, IGFBP-6, IGF-1 R, livin, p21, p27, p53, SMAC, Survivin (BIRC5), TNF RI (TNFRSF1A), TNF RII (TNFRSF1B), TNF alpha, TNF beta (TNFSF1B), TRAIL R1 (TNFRSF10A/DR4), TRAIL R2 (TNFRSF10B/DR5), TRAIL R3 (TNFRSF10C), TRAIL R4 (TNFRSF10D), XIAP
Catalog No. 50-194-8477 Supplier Raybiotech Inc Supplier No. AAHAPO18
Detects 43 Human Apoptotic Factors. Suitable for all liquid sample types but intended for use with cell and tissue lysates. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: bad, bax, bcl-2, bcl-w, BID, BIM, Caspase-3, Caspase-8, CD40 (TNFRSF5), CD40 Ligand (TNFSF5), cIAP-2, Cytochrome C, DR6 (TNFRSF21), Fas (TNFRSF6/Apo-1), Fas Ligand (TNFSF6), HSP27, HSP60, HSP70, HTRA2, IGF-1, IGF-2, IGFBP-1, IGFBP-2, IGFBP-3, IGFBP-4, IGFBP-5, IGFBP-6, IGF-1 R, livin, p21, p27, p53, SMAC, Survivin (BIRC5), TNF RI (TNFRSF1A), TNF RII (TNFRSF1B), TNF alpha, TNF beta (TNFSF1B), TRAIL R1 (TNFRSF10A/DR4), TRAIL R2 (TNFRSF10B/DR5), TRAIL R3 (TNFRSF10C), TRAIL R4 (TNFRSF10D), XIAP