Learn More
Biomatik Corporation Human Apoptosis Inducing Factor (AIF) CLIA Kit
Supplier:  Biomatik Corporation EKU08931
Encompass_Preferred
Human Apoptosis Inducing Factor (AIF) CLIA Kit for detecting AIF in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 2.74-2,000pg/ml. Sensitivity: 1.17pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: O95831. Target Synonyms: PDCD8; Programmed Cell Death protein 8; Apoptosis-inducing factor 1, mitochondrial. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8590