Learn More
American Research Products Inc Human apoptosis inducing facto
Catalog No. 5000329245
Encompass
Human apoptosis inducing factor AIF ELISA Kit from CUSABIO Detection Range 23 5 pg/mL-1500 pg/mL Sensitivity 5 8 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-29245 Supplier American Research Products Inc Supplier No. CSBE09006H
Human apoptosis inducing factor AIF ELISA Kit from CUSABIO Detection Range 23 5 pg/mL-1500 pg/mL Sensitivity 5 8 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich