Learn More
Biomatik Corporation Human Apoptosis Inhibitor 5 (API5) ELISA Kit
Supplier:  Biomatik Corporation EKU08492
Encompass_Preferred
Human Apoptosis Inhibitor 5 (API5) ELISA Kit for detecting API5 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.121ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9BZZ5. Gene ID: 8539. Target Synonyms: MIG8; FIF; AAC11; API5L1; XAGL; Cell migration-inducing gene 8; Antiapoptosis clone 11; Fibroblast Growth Factor 2-Interacting Factor 2; Migration-Inducing Protein MIG8. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5611