Learn More
American Research Products Inc Human Apoptosis regulator BAX
Catalog No. 5000326178
Encompass
Human Apoptosis regulator BAX ELISA Kit from CUSABIO Detection Range 1 25 ng/mL-80 ng/mL Sensitivity 0 312 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26178 Supplier American Research Products Inc Supplier No. CSBE09344H
Human Apoptosis regulator BAX ELISA Kit from CUSABIO Detection Range 1 25 ng/mL-80 ng/mL Sensitivity 0 312 ng/mL Sample serum plasma tissue homogenates Method Sandwich