Learn More
Biomatik Corporation Human Apotransferrin (apoTf) CLIA Kit
Supplier:  Biomatik Corporation EKU08948
Encompass_Preferred
Human Apotransferrin (apoTf) CLIA Kit for detecting apoTf in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 27.4-20,000pg/ml. Sensitivity: 12.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P02787. Gene ID: 7018. Target Synonyms: apo-Transferrin; Apo-Tf. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6799