Learn More
Sino Biological Anti-Beta-amyloid 40 / Beta-APP40 Antibody
Supplier:  Sino Biological 10703RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Beta-Amyloid / Abeta / APP (rh Beta-Amyloid / Abeta / APP; Catalog#10703-H20E1; P05067-1; Asp672-Val711). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-1464