Learn More
Sino Biological Recombinant Human Beta-amyloid/Beta-APP Protein (hFc Tag)
Supplier:  Sino Biological 10703H02H50
Encompass_Preferred
A DNA sequence encoding the human APP-751 isoform (NP_958816.1) (Met 1-Leu 669) was expressed with the C-terminal fused Fc region of human IgG1.
Catalog No. 50-224-6939