Learn More
ACROBiosystems Human APRIL 25ug
Supplier:  ACROBiosystems APLH82Q825UG
Encompass_Preferred
Biotinylated Human APRIL Protein AvitagHis Tag (APL-H82Q8) is expressed from human 293 cells (HEK293). It contains AA Gln 111 - Leu 250 (Accession # O75888-1).
Catalog No. 50-253-5495