Learn More
ACROBiosystems Human APRIL Protein Fc Tag
Supplier:  ACROBiosystems APLH5263100UG
Encompass_Preferred
Human APRIL Protein Fc Tag (APL-H5263) is expressed from human 293 cells (HEK293). It contains AA Gln 111 - Leu 250 (Accession # O75888-1).
Catalog No. 50-253-5497