Learn More
Gibco™ Human APRIL Recombinant Protein, PeproTech®
Description
310-10C-01M will be provided as 20 x 50 μg (310-10C-50UG). 310-10C-500 will be provided as 10 x 50 μg (310-10C-50UG). 310-10C-250 will be provided as 5 x 50 μg (310-10C-50UG). 310-10C-100 will be provided as 2 x 50 μg (310-10C-50UG) Recombinant human APRIL is a soluble 16.3 kDa protein, consisting of 146 amino acid residues. *Human APRIL (Catalog Number 310-10C) has replaced Human APRIL (Catalog Number 310-10). This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis.
Specifications
Specifications
| Accession Number | O75888 |
| For Use With (Application) | Control, Bioactivity |
| Formulation | Protein with no preservative |
| Gene ID (Entrez) | 8741 |
| Molecular Weight (g/mol) | 16.3 kDa |
| Name | Human APRIL |
| Purification Method | Purified |
| Quantity | 50 μg |
| Source | Insect cells |
| Storage Requirements | -20°C |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.